Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Enpep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007934.3 | NP_031960.1 | 20 | 3301 | ||
GenBank mRNA | AK031439.1 | - | 19 | 2333 | ||
AK128975.1 | - | 20 | 3258 | |||
AK146808.1 | - | 20 | 3273 | |||
BC064061.1 | - | 5 | 1034 | |||
M29961.1 | - | 20 | 3120 |