Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ereg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007950.2 | NP_031976.1 | 5 | 2244 | ||
GenBank mRNA | D30782.1 | - | 5 | 2243 |