Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007963.2 | NP_031989.2 | 13 | 2567 | ||
GenBank mRNA | AJ001482.1 | - | 12 | 2722 | ||
AK033517.1 | - | 13 | 2001 | |||
BC076620.1 | - | 13 | 2519 | |||
BC139762.1 | - | 11 | 1685 | 40109903 | ||
JQ665270.1 | - | 11 | 2687 |