Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Evi5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007964.2 | NP_031990.2 | 6 | 862 | ||
GenBank mRNA | AK134294.1 | - | 6 | 1063 | ||
AK164899.1 | - | 3 | 187 | |||
AK165332.1 | - | 7 | 855 | |||
AK165885.1 | - | 6 | 815 | |||
AK192278.1 | - | 4 | 352 | |||
BC030056.1 | - | 7 | 869 | |||
U53586.1 | - | 6 | 862 |