Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fabp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007980.3 | NP_032006.1 | 4 | 742 | ||
GenBank mRNA | AK007995.1 | - | 4 | 440 | ||
AY523818.1 | - | 4 | 428 | |||
AY523819.1 | - | 4 | 428 | |||
BC013457.1 | - | 4 | 422 | 4218050 | ||
BY707992.1 | - | 4 | 438 | |||
M65034.1 | - | 4 | 436 |