Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007986.3 | NP_032012.1 | 14, 15 | 1393 | ||
GenBank mRNA | AK136578.1 | - | 13, 14 | 1383 | ||
BC019190.1 | - | 14, 15 | 1297 | 5028579 | ||
Y10007.1 | - | 14, 15 | 1393 |