Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008023.1 | NP_032049.1 | 1 | 982 | ||
GenBank mRNA | BC115976.1 | - | 2 | 672 | ||
BC139476.1 | - | 1 | 957 | 8862498 | ||
BC139477.1 | - | 1 | 957 | 8862499 | ||
X92591.1 | - | 1 | 982 |