Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fstl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008047.5 | NP_032073.2 | 4 | 373 | ||
GenBank mRNA | AK049440.1 | - | 4 | 374 | ||
AK147117.1 | - | 4 | 371 | |||
BC006185.1 | - | 4 | 401 | |||
BC028921.1 | - | 4 | 357 | 2647002 | ||
M91380.1 | - | 4 | 354 |