Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008051.6 | NP_032077.2 | 2 | 243 | ||
GenBank mRNA | AF113533.1 | - | 1 | 99 | ||
AK137638.1 | - | 2 | 245 | |||
BY025357.1 | - | 2 | 233 |