Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293621.1 | NP_001280550.1 | 4 | 349 | ||
NM_008060.2 | NP_032086.1 | 4 | 349 | |||
GenBank mRNA | AK028655.1 | - | 4 | 317 | ||
AK030722.1 | - | 4 | 318 | |||
AK043606.1 | - | 4 | 296 | |||
AK081915.1 | - | 4 | 311 | |||
AK149686.1 | - | 4 | 306 | |||
AK166150.1 | - | 4 | 288 | |||
BC094437.1 | - | 4 | 299 | 30682580 | ||
BC117888.1 | - | 4 | 292 | 40096413 | ||
BC117889.1 | - | 4 | 295 | 40096414 | ||
BC128069.1 | - | 4 | 293 | 40096428 | ||
U92793.1 | - | 4 | 295 |