Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gata5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008093.2 | NP_032119.2 | 4 | 1172 | ||
GenBank mRNA | AK132020.1 | - | 4 | 1193 | ||
AK142213.1 | - | 4 | 1174 | |||
BC105653.1 | - | 5 | 885 | 40046623 | ||
BC105654.1 | - | 5 | 946 | 40046622 | ||
BC105655.1 | - | 5 | 795 | 40046621 | ||
BC105656.1 | - | 5 | 451 | |||
U84725.1 | - | 4 | 1166 |