Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024501.2 | NP_078777.1 | 1 | 866 | ||
GenBank mRNA | AF241528.1 | - | 1 | 866 | ||
AK314120.1 | - | 1 | 781 | |||
BC014477.1 | - | 1 | 770 | 4869019 |