Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008027.2 | NP_032053.1 | 5, 6 | 521 | ||
GenBank mRNA | AF145044.1 | - | 4, 5 | 362 | ||
AK150692.1 | - | 5, 6 | 543 | |||
AK150827.1 | - | 5, 6 | 470 | |||
AK167323.1 | - | 5, 6 | 448 | |||
AK181092.1 | - | 2, 3 | 191 | |||
AY167925.1 | - | 5, 6 | 527 | |||
BC004647.1 | - | 5, 6 | 531 | 3158071 | ||
U90435.1 | - | 5, 6 | 521 |