Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fmr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290424.1 | NP_001277353.1 | 4, 5 | 538 | ||
NM_008031.3 | NP_032057.2 | 4, 5 | 538 | |||
GenBank mRNA | AF461114.1 | - | 4, 5 | 262 | ||
AK046605.1 | - | 4, 5 | 498 | |||
AK053211.1 | - | 1, 2 | 280 | |||
AK053701.1 | - | 4, 5 | 498 | |||
AK053829.1 | - | 4, 5 | 540 | |||
AK080948.1 | - | 4, 5 | 498 | |||
AK140343.1 | - | 4, 5 | 500 | |||
AK208300.1 | - | 4, 5 | 238 | |||
BC079671.1 | - | 4, 5 | 524 | 30532682 | ||
L23971.1 | - | 4, 5 | 385 |