Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Folr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252552.1 | NP_001239481.1 | 4 | 645 | ||
NM_001252553.1 | NP_001239482.1 | 4 | 571 | |||
NM_001252554.1 | NP_001239483.1 | 4 | 558 | |||
NM_008034.3 | NP_032060.2 | 4 | 642 | |||
GenBank mRNA | AF096319.1 | - | 4 | 537 | ||
AK002275.1 | - | 4 | 639 | |||
BC002071.1 | - | 1 | 211 | |||
BC023477.1 | - | 1 | 192 | |||
BC037078.1 | - | 3 | 383 | |||
BC138781.1 | - | 3 | 275 | 8861803 | ||
BC138795.1 | - | 3 | 362 | 8861817 | ||
BC145414.1 | - | 3 | 362 | 9053972 | ||
CF617679.1 | - | 3 | 388 | |||
M64782.1 | - | 1 | 211 |