Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271981.1 | NP_001258910.1 | 2 | 279 | ||
NM_008051.6 | NP_032077.2 | 2 | 417 | |||
GenBank mRNA | AF113533.1 | - | 1 | 273 | ||
AK137638.1 | - | 2 | 419 | |||
BC109145.1 | - | 2 | 255 | 40045410 | ||
BC109146.1 | - | 2 | 255 | 40045419 |