Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PBX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025245.2 | NP_079521.1 | 2, 3 | 478 | ||
NR_038198.1 | 2, 3 | 478 | ||||
GenBank mRNA | AJ300182.1 | - | 1, 2 | 59 | ||
AK097427.1 | - | 2, 3 | 215 | |||
AK301631.1 | - | 2, 3 | 243 | |||
BC141859.1 | - | 2, 3 | 478 | 8860384 | ||
BC143588.1 | - | 2, 3 | 254 | |||
BC143589.1 | - | 2, 3 | 255 | |||
BG683004.1 | - | 2, 3 | 193 |