Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008067.4 | NP_032093.3 | 7, 8 | 1061 | ||
GenBank mRNA | AK039144.1 | - | 7, 8 | 1002 | ||
AK134760.1 | - | 7, 8 | 970 | |||
BC119317.1 | - | 7, 8 | 859 | 8734066 | ||
BC120686.1 | - | 7, 8 | 859 | 40129609 | ||
M86568.1 | - | 7, 8 | 963 |