Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001038701.2 | NP_001033790.1 | 4 | 346 | ||
NM_008071.3 | NP_032097.1 | 4 | 398 | |||
GenBank mRNA | AK039070.1 | - | 4 | 368 | ||
AK083051.1 | - | 4 | 410 | |||
AK083107.1 | - | 4 | 347 | |||
AK147247.1 | - | 4 | 352 | |||
U14420.1 | - | 4 | 379 |