Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gabrr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008076.3 | NP_032102.2 | 4, 5 | 662 | ||
GenBank mRNA | AF024621.1 | - | 4, 5 | 775 | ||
AK132642.1 | - | 4, 5 | 490 | |||
AK160446.1 | - | 2, 3 | 329 | |||
BC057957.1 | - | 4, 5 | 647 | 5357713 |