Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gas1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008086.2 | NP_032112.1 | 1 | 2078 | ||
GenBank mRNA | BC055747.1 | - | 2 | 1173 | ||
BC058628.1 | - | 2 | 1718 | 6401957 | ||
X65128.1 | - | 1 | 1908 |