Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcgr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008101.2 | NP_032127.2 | 8 | 977 | ||
GenBank mRNA | AK004890.1 | - | 5 | 1184 | ||
AK144393.1 | - | 8 | 924 | |||
BC012718.1 | - | 7 | 1052 | |||
BC031885.1 | - | 8 | 927 | 5137340 | ||
BC057988.1 | - | 8 | 908 | 5100574 | ||
L38613.1 | - | 7 | 887 |
Set Membership: |
G-protein coupled receptor |