Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HMGN5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030763.2 | NP_110390.1 | 4, 5 | 402 | ||
XM_017029840.1 | XP_016885329.1 | 2, 3 | 226 | |||
GenBank mRNA | AF250329.1 | - | 3, 4 | 141 | ||
AK094058.1 | - | 4, 5 | 402 | |||
AK315792.1 | - | 3, 4 | 195 | |||
BC005342.1 | - | 3, 4 | 162 | 3838879 | ||
DA745749.1 | - | 4, 5 | 371 |