Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnta | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008033.3 | NP_032059.1 | 9 | 1073 | ||
GenBank mRNA | AK010744.1 | - | 9 | 1074 | ||
AK090346.1 | - | 9 | 1073 | |||
AK153252.1 | - | 9 | 1068 | |||
BC012711.1 | - | 9 | 1091 | 4019240 | ||
D49744.1 | - | 9 | 1067 |