Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162494.1 | NP_001155966.1 | 7 | 3926 | ||
NM_008056.3 | NP_032082.3 | 7 | 3930 | |||
GenBank mRNA | AK088399.1 | - | 7 | 3795 | ||
AK138964.1 | - | 4 | 2643 | |||
BC026150.1 | - | 7 | 3919 | 3983985 |
Set Membership: |
G-protein coupled receptor |