Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ganab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293621.1 | NP_001280550.1 | 24 | 3408 | ||
NM_008060.2 | NP_032086.1 | 25 | 3474 | |||
GenBank mRNA | AK017873.1 | - | 12 | 1814 | ||
AK028655.1 | - | 24 | 3378 | |||
AK030722.1 | - | 24 | 3377 | |||
AK081915.1 | - | 24 | 3370 | |||
AK166150.1 | - | 24 | 3342 | |||
AK213427.1 | - | 1 | 207 | |||
BC020386.1 | - | 17 | 2624 | |||
BC035423.1 | - | 15 | 2224 | |||
BC094437.1 | - | 24 | 3358 | 30682580 | ||
U92793.1 | - | 25 | 3420 |