Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Galc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK154760.1 | - | 15 | 1889 | ||
AK161339.1 | - | 10 | 1179 | |||
BC021641.1 | - | 2 | 146 | |||
BC065140.1 | - | 15 | 1761 | |||
BC086671.1 | - | 15 | 1766 | 30635559 | ||
D38557.1 | - | 15 | 1836 |