Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PIP4K2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005028.4 | NP_005019.2 | 7 | 1015 | ||
XM_006717450.2 | XP_006717513.1 | 6 | 857 | |||
XM_011519496.1 | XP_011517798.1 | 7 | 791 | |||
XM_017016330.1 | XP_016871819.1 | 7 | 849 | |||
XM_017016331.1 | XP_016871820.1 | 7 | 944 | |||
XM_017016332.1 | XP_016871821.1 | 6 | 657 | |||
GenBank mRNA | AK294817.1 | - | 7 | 796 | ||
AK294867.1 | - | 5 | 402 | |||
BC018034.1 | - | 7 | 996 | 4817357 | ||
S78798.1 | - | 7 | 1081 | |||
U14957.2 | - | 7 | 789 |