Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Grb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008163.3 | NP_032189.1 | 3 | 453 | ||
GenBank mRNA | AK077750.1 | - | 2 | 272 | ||
AK079087.1 | - | 2 | 280 | |||
AK148148.1 | - | 3 | 418 | |||
AK153557.1 | - | 2 | 386 | |||
AK155800.1 | - | 3 | 408 | |||
AK166142.1 | - | 3 | 407 | |||
AK170845.1 | - | 2 | 244 | |||
AK172002.1 | - | 3 | 428 | |||
BC052377.1 | - | 3 | 453 | 5696248 | ||
BC085254.1 | - | 3 | 289 | 5343274 | ||
D85748.1 | - | 2 | 200 | |||
U07617.1 | - | 2 | 202 |