Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113325.2 | NP_001106796.1 | 2 | 535 | ||
NM_001252403.1 | NP_001239332.1 | 2 | 162 | |||
NM_008165.4 | NP_032191.2 | 2 | 535 | |||
GenBank mRNA | AB102773.1 | - | 2 | 119 | ||
AB102774.1 | - | 2 | 119 | |||
AK049958.1 | - | 2 | 345 | |||
AK135107.1 | - | 2 | 163 | |||
BC056397.1 | - | 2 | 197 | 6842391 | ||
BC060702.1 | - | 2 | 118 | |||
CJ155313.1 | - | 2 | 463 | |||
JN180918.1 | - | 186 | ||||
JN180919.1 | - | 2 | 117 | |||
X57497.1 | - | 2 | 382 |