Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gm3776 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243092.1 | NP_001230021.1 | 7 | 755 | ||
GenBank mRNA | BC061134.1 | - | 725 | 30308304 |
Gene Symbol: Gsta1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008181.3 | NP_032207.3 | 7 | 742 | ||
GenBank mRNA | BC132572.1 | - | 725 | 40130849 | ||
BC132576.1 | - | 725 | 40130853 | |||
CJ228212.1 | - | 5 | 599 |