Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H3f3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008210.4 | NP_032236.1 | 3 | 395 | ||
GenBank mRNA | AK008405.1 | - | 2 | 296 | ||
AK037900.1 | - | 3 | 397 | |||
AK150467.1 | - | 3 | 355 | |||
AK150468.1 | - | 3 | 355 | |||
AK150591.1 | - | 3 | 350 | |||
AK150928.1 | - | 3 | 350 | |||
AK151634.1 | - | 3 | 355 | |||
AK152293.1 | - | 3 | 350 | |||
AK153034.1 | - | 3 | 354 | |||
AK160635.1 | - | 3 | 350 | |||
AK160677.1 | - | 3 | 354 | |||
AK162040.1 | - | 3 | 353 | |||
AK167879.1 | - | 3 | 354 | |||
AK168197.1 | - | 3 | 354 | |||
AK169608.1 | - | 3 | 350 | |||
AK205408.1 | - | 1 | 50 | |||
AK215513.1 | - | 2 | 165 | |||
AK217933.1 | - | 2 | 165 | |||
BC002268.1 | - | 3 | 340 | 3495830 | ||
BC012687.1 | - | 3 | 344 | 4224280 | ||
BC083353.1 | - | 3 | 335 | 5714859 | ||
BC088835.1 | - | 3 | 332 | 6437687 | ||
BC106177.1 | - | 3 | 329 | 6511787 | ||
Z85979.1 | - | 3 | 343 |
Gene Symbol: Gm12657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081019.1 | NP_001074488.1 | 2 | 282 |