Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008224.4 | NP_032250.2 | 1 | 488 | ||
GenBank mRNA | AK088827.1 | - | 1 | 487 | ||
AK140197.1 | - | 1 | 474 | |||
BC053742.1 | - | 1 | 446 | 6509535 |