Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hdc | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008230.6 | NP_032256.3 | 2 | 221 | ||
GenBank mRNA | AF109137.1 | - | 2 | 131 | ||
AK076384.1 | - | 2 | 205 | |||
AK088545.1 | - | 2 | 208 | |||
AK133455.1 | - | 2 | 222 | |||
AK150168.1 | - | 2 | 209 | |||
AK153104.1 | - | 2 | 209 | |||
BC052833.1 | - | 2 | 206 | 30044098 | ||
X57437.1 | - | 2 | 186 |