Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gnai2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008138.4 | NP_032164.2 | 4 | 478 | ||
GenBank mRNA | AK157998.1 | - | 4 | 482 | ||
AK159222.1 | - | 4 | 482 | |||
AK160186.1 | - | 4 | 474 | |||
AK167388.1 | - | 4 | 480 | |||
AK185434.1 | - | 1 | 52 | |||
AK185480.1 | - | 377 | ||||
AK202003.1 | - | 4 | 358 | |||
BC065159.1 | - | 4 | 465 | 5717305 | ||
M13963.1 | - | 4 | 450 | |||
S79607.1 | - | 3 | 235 |