Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167694.2 | NP_001161166.1 | 4 | 869 | ||
NM_001167695.2 | NP_001161167.1 | 4 | 800 | |||
NM_001167696.2 | NP_001161168.1 | 4 | 830 | |||
NM_001167697.2 | NP_001161169.1 | 3 | 917 | |||
NM_001167699.2 | NP_001161171.1 | 2 | 799 | |||
NM_001167700.2 | NP_001161172.1 | 5 | 1019 | |||
NM_008157.3 | NP_032183.2 | 2 | 760 | |||
GenBank mRNA | AK132917.1 | - | 5 | 958 | ||
AK140770.1 | - | 5 | 1215 | |||
BC021648.1 | - | 4 | 808 | 3989077 | ||
BC050187.1 | - | 3 | 888 | 30020126 | ||
U46923.1 | - | 2 | 620 |
Set Membership: |
G-protein coupled receptor |