Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ABCC11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032583.3 | NP_115972.2 | 16, 17 | 2277 | ||
NM_033151.3 | NP_149163.2 | 15, 16 | 2563 | |||
NM_145186.2 | NP_660187.1 | 16, 17 | 2277 | |||
XM_011523397.1 | XP_011521699.1 | 12, 13 | 1783 | |||
XM_011523398.2 | XP_011521700.1 | 4, 5 | 498 | |||
XM_017023795.1 | XP_016879284.1 | 16, 17 | 4891 | |||
XM_017023796.1 | XP_016879285.1 | 17, 18 | 2364 | |||
XM_017023797.1 | XP_016879286.1 | 17, 18 | 2492 | |||
XM_017023798.1 | XP_016879287.1 | 16, 17 | 2318 | |||
XM_017023799.1 | XP_016879288.1 | 16, 17 | 2300 | |||
XM_017023800.1 | XP_016879289.1 | 16, 17 | 2291 | |||
XM_017023801.1 | XP_016879290.1 | 15, 16 | 4783 | |||
XM_017023802.1 | XP_016879291.1 | 10, 11 | 1638 | |||
XM_017023803.1 | XP_016879292.1 | 16, 17 | 4891 | |||
GenBank mRNA | AF352582.1 | - | 16, 17 | 2312 | ||
AF367202.1 | - | 16, 17 | 2291 | |||
AF411579.1 | - | 16, 17 | 2291 | |||
AY040219.1 | - | 15, 16 | 2563 |