Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gucy2e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008192.3 | NP_032218.2 | 6, 7 | 1669 | ||
GenBank mRNA | AK044282.1 | - | 6, 7 | 1670 | ||
BC057954.1 | - | 6, 7 | 1654 |