Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: H1f0 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008197.3 | NP_032223.2 | 1 | 854 | ||
GenBank mRNA | AK051213.1 | - | 1 | 617 | ||
AK077473.1 | - | 1 | 531 | |||
AK090029.1 | - | 1 | 619 | |||
AK145675.1 | - | 1 | 506 | |||
AK146077.1 | - | 1 | 617 | |||
AK153990.1 | - | 1 | 618 | |||
AK155312.1 | - | 1 | 549 | |||
AK177351.1 | - | 1 | 112 | |||
AK186840.1 | - | 1 | 112 | |||
BC003830.1 | - | 1 | 603 | 3600538 | ||
BC011493.1 | - | 1 | 601 | 4166167 | ||
BC110361.1 | - | 1 | 606 | 5343233 | ||
X13171.1 | - | 1 | 584 |