Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hadh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008212.4 | NP_032238.2 | 2 | 315 | ||
GenBank mRNA | AK075609.1 | - | 2 | 242 | ||
AK132260.1 | - | 2 | 278 | |||
AK148481.1 | - | 2 | 239 | |||
AK148486.1 | - | 2 | 252 | |||
AK167024.1 | - | 2 | 242 | |||
AK168238.1 | - | 2 | 262 | |||
AK168877.1 | - | 2 | 242 | |||
AK169261.1 | - | 2 | 281 | |||
BC028833.1 | - | 2 | 147 | |||
BC064712.1 | - | 2 | 257 | 30303218 | ||
D29639.1 | - | 2 | 241 |