Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Has1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008215.2 | NP_032241.1 | 2 | 716 | ||
GenBank mRNA | AK053726.1 | - | 2 | 735 | ||
BC125424.1 | - | 2 | 695 | 40129939 | ||
BC125426.1 | - | 2 | 695 | 40129941 | ||
D82964.1 | - | 2 | 722 |