Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LMNB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032737.3 | NP_116126.3 | 5 | 793 | ||
GenBank mRNA | AL560944.3 | - | 792 | |||
BC006551.2 | - | 5 | 758 | 2961134 | ||
BT007441.1 | - | 5 | 698 | |||
KP137589.1 | - | 2 | 347 | |||
M94362.1 | - | 4 | 446 |