Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcls1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008225.2 | NP_032251.2 | 6 | 524 | ||
GenBank mRNA | AK088723.1 | - | 6 | 506 | ||
BC007469.1 | - | 6 | 494 | 3593456 | ||
D42120.1 | - | 6 | 455 | |||
X84797.1 | - | 6 | 492 |