Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CASP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001224.4 | NP_001215.1 | 2 | 192 | ||
NM_032982.3 | NP_116764.2 | 2 | 438 | |||
NM_032983.3 | NP_116765.2 | 2 | 438 | |||
GenBank mRNA | AF314174.1 | - | 2 | 146 | ||
AF314175.1 | - | 1 | 104 | |||
AK291274.1 | - | 2 | 230 | |||
AK303391.1 | - | 2 | 210 | |||
BC002427.2 | - | 2 | 210 | 3346279 | ||
BG397039.1 | - | 2 | 269 | |||
BT007240.1 | - | 2 | 146 | |||
CB988674.1 | - | 2 | 371 | |||
CR541748.1 | - | 1 | 104 | |||
DA505883.1 | - | 2 | 439 | |||
DB455837.1 | - | 2 | 403 | |||
DB455840.1 | - | 2 | 403 | |||
KU177957.1 | - | 2 | 197 | |||
KU177958.1 | - | 2 | 197 | |||
U13021.1 | - | 2 | 159 | |||
U13022.1 | - | 2 | 192 |