Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037741.3 | NP_001032830.2 | 4 | 537 | ||
NM_008162.3 | NP_032188.3 | 4 | 483 | |||
GenBank mRNA | AB072797.1 | - | 4 | 519 | ||
AF045768.1 | - | 4 | 401 | |||
AF045769.1 | - | 4 | 292 | |||
AF274027.1 | - | 4 | 513 | |||
AK006441.1 | - | 4 | 378 | |||
AK145770.1 | - | 4 | 403 | |||
AK160389.1 | - | 4 | 310 | |||
AK160509.1 | - | 4 | 286 | |||
AK167881.1 | - | 4, 5 | 387 | |||
AK168027.1 | - | 4 | 310 | |||
BC083137.1 | - | 3 | 270 | |||
BC106147.1 | - | 4 | 330 | 4936866 | ||
BY238083.1 | - | 4 | 407 | |||
CA466677.1 | - | 4 | 464 | |||
D87896.1 | - | 4 | 483 |