Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gria1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113325.2 | NP_001106796.1 | 15, 16 | 2944 | ||
NM_001252403.1 | NP_001239332.1 | 15, 16 | 2571 | |||
NM_008165.4 | NP_032191.2 | 15, 16 | 2944 | |||
GenBank mRNA | AB102773.1 | - | 15, 16 | 2528 | ||
AB102774.1 | - | 15, 16 | 2528 | |||
AB104707.1 | - | 4, 5 | 801 | |||
AK013839.1 | - | 2, 3 | 220 | |||
AK049958.1 | - | 15, 16 | 2754 | |||
AK135107.1 | - | 15, 16 | 2572 | |||
BC056397.1 | - | 15, 16 | 2606 | 6842391 | ||
BC060702.1 | - | 15, 16 | 2527 | |||
X57497.1 | - | 15, 16 | 2791 |