Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guca2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008191.2 | NP_032217.2 | 3 | 537 | ||
GenBank mRNA | AK002364.1 | - | 3 | 509 | ||
AV002130.1 | - | 2 | 290 | |||
AV003693.1 | - | 1 | 169 | |||
U67800.1 | - | 3 | 493 | |||
U90727.1 | - | 3 | 539 |