Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Guk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159410.1 | NP_001152882.1 | 7, 8 | 610 | ||
NM_008193.3 | NP_032219.2 | 7, 8 | 773 | |||
GenBank mRNA | AK004205.1 | - | 7, 8 | 632 | ||
AK013514.1 | - | 7, 8 | 636 | |||
BC024625.1 | - | 7, 8 | 637 | 4485215 | ||
U53514.1 | - | 7, 8 | 593 |