Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hcfc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008224.4 | NP_032250.2 | 3, 4 | 865 | ||
GenBank mRNA | AJ627036.1 | - | 31 | |||
AK088827.1 | - | 3, 4 | 864 | |||
AK140197.1 | - | 3, 4 | 851 | |||
BC053742.1 | - | 3, 4 | 823 | 6509535 |