Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hmgb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293623.1 | NP_001280552.1 | 2 | 405 | ||
NM_001293624.1 | NP_001280553.1 | 2 | 176 | |||
NM_001293625.1 | NP_001280554.1 | 2 | 172 | |||
NM_008253.4 | NP_032279.1 | 2 | 358 | |||
GenBank mRNA | AF022465.1 | - | 1 | 106 | ||
AI385779.1 | - | 2 | 358 | |||
AK032163.1 | - | 2 | 282 | |||
AK141399.1 | - | 2 | 175 | |||
AK146795.1 | - | 2 | 175 | |||
AK159119.1 | - | 2 | 149 | |||
AK159322.1 | - | 2 | 149 | |||
BC011276.1 | - | 2 | 139 | 3980194 | ||
BC083352.1 | - | 1 | 90 | 30540227 | ||
BC158037.1 | - | 2 | 110 | 9007340 | ||
CX240626.1 | - | 2 | 141 |